5-Bromo-2-nitrophenol 96%
Supplier: Thermo Scientific Chemicals
|
Warning
|
H64966-06
H64966-14
AAH64966-06EA
68.5
USD
AAH64966-06
AAH64966-14
5-Bromo-2-nitrophenol 96%
5-Bromo-2-nitrophenol
Purity: 96%
CAS No.: 27684-84-0
Molecular Formula: C6H4BrNO3
Formula Weight: 218.01
Physical Form: Crystalline solid
Appearance: Pale yellow
Melting Point: 40-41°
Beilstein Reference: 2329085
MDL No.: MFCD03095027
Formula:
C₆H₄BrNO₃ Melting Pt: 40…41 °C Storage Temperature: Ambient |
MDL Number:
MFCD03095027 CAS Number: 27684-84-0 |
Specification Test Results
Assay Minimum | 96 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...