3-Aminopyridazine 97%

Supplier: Thermo Scientific Chemicals
Warning

Synonyms: Pyridazin-3-amine

H64859-03 H64859-06 H64859-14
AAH64859-03EA 36.8 USD
AAH64859-03 AAH64859-06 AAH64859-14
3-Aminopyridazine 97%
3-Aminopyridazine

Purity: 97% CAS No.: 5469-70-5 Molecular Formula: C4H5N3 Formula Weight: 95.10 Physical Form: Solid Appearance: White to pale yellow Melting Point: 168-172° Beilstein Reference: 107024 MDL No.: MFCD01529869

Formula: C4H5N3
MW: 95.10 g/mol
Melting Pt: 168…172 °C
Storage Temperature: Ambient
MDL Number: MFCD01529869
CAS Number: 5469-70-5

Order Now

Specification Test Results

Assay Minimum 97


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR