2-Methylpropanethioamide 95%

Supplier: Thermo Scientific Chemicals
Warning

Synonyms: Thioisobutyramide

H64663-03 H64663-06 H64663-14
AAH64663-03EA 47.8 USD
AAH64663-03 AAH64663-06 AAH64663-14
2-Methylpropanethioamide 95%
2-Methylpropanethioamide

Purity: 95% CAS No.: 13515-65-6 Molecular Formula: C4H9NS Formula Weight: 103.18 Storage Temperature: -30°C to -10°C Physical Form: Solid Melting Point: 45-48° Beilstein Reference: 1098427 MDL No.: MFCD07369538

Formula: C₄H₉NS
Melting Pt: 45…48 °C
Storage Temperature: Refrigerator
MDL Number: MFCD07369538
CAS Number: 13515-65-6

Order Now

Specification Test Results

Assay Minimum 95


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR