L-isoleucine ethyl ester hydrochloride 98%

Supplier: Thermo Scientific Chemicals

H64640-06 H64640-14
AAH64640-03EA 24.13 USD
AAH64640-03 AAH64640-06 AAH64640-14
L-isoleucine ethyl ester hydrochloride 98%
L-isoleucine ethyl ester hydrochloride

Purity: 98% CAS No.: 56782-52-6 Molecular Formula: C8H17NO2.HCl Formula Weight: 195.69 Physical Form: Powder Appearance: White Beilstein Reference: 4710859 MDL No.: MFCD09953729

Formula: C₈H₁₈ClNO₂
Storage Temperature: Ambient
MDL Number: MFCD09953729
CAS Number: 56782-52-6
EINECS: 260-382-1

Order Now

Specification Test Results

Assay Minimum 98

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR