2-Bromo-5-chlorobenzaldehyde 97%
Supplier: Thermo Scientific Chemicals
|
Warning
|
H64330-06
H64330-14
H64330-22
AAH64330-06EA
28.8
USD
AAH64330-06
AAH64330-14
AAH64330-22
2-Bromo-5-chlorobenzaldehyde 97%
2-Bromo-5-chlorobenzaldehyde
Purity: 97%
CAS No.: 174265-12-4
Molecular Formula: C7H4BrClO
Formula Weight: 219.46
Physical Form: Crystalline
Appearance: Pale yellow
Melting Point: 64-68° lit.
Beilstein Reference: 10423891
MDL No.: MFCD00462870
Formula:
C₇H₄BrClO Melting Pt: 64…68 °C (lit.) Storage Temperature: Ambient |
MDL Number:
MFCD00462870 CAS Number: 174265-12-4 |
Specification Test Results
Assay Minimum | 97 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...