3-Cyano-4-fluorophenylboronic acid pinacol ester 96%
Supplier: Thermo Scientific Chemicals
|
Danger
|
H62245-03
H62245-06
AAH62245-03EA
74.92
USD
AAH62245-03
AAH62245-06
AAH62245-MD
3-Cyano-4-fluorophenylboronic acid pinacol ester 96%
3-Cyano-4-fluorophenylboronic acid pinacol ester
Purity: 96%
CAS No.: 775351-57-0
Molecular Formula: C13H15BFNO2
Formula Weight: 247.07
Physical Form: Solid
Melting Point: 75-76°
MDL No.: MFCD06795681
Formula:
C₁₃H₁₅BFNO₂ Melting Pt: 75…76 °C Storage Temperature: Ambient |
MDL Number:
MFCD06795681 CAS Number: 775351-57-0 UN: 3439 ADR: "6.1",III |
Specification Test Results
Assay Minimum | 96 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...