3-Methyl-2-buten-1-ol 97%
Supplier: Thermo Scientific Chemicals
|
Danger
|
Synonyms:
2-Methyl-2-buten-4-ol
H36436-36
H36436-22
AAH36436-36EA
90.48
USD
AAH36436-36
AAH36436-22
3-Methyl-2-buten-1-ol 97%
3-Methyl-2-buten-1-ol
Formula:
C₅H₁₀O MW: 86.13 g/mol Boiling Pt: 139…140°C Density: 0.86 Flash Pt: 43 °C(109 °F) Storage Temperature: Ambient |
MDL Number:
MFCD00002916 CAS Number: 556-82-1 EINECS: 209-141-4 UN: 1987 ADR: "3",III |
Specification Test Results
Assay Minimum | 97 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...