Tetrabutylammonium hydroxide 55% w/w water
Supplier: BeanTown Chemical
225850-1KG
225850-250G
225850-50G
BT225850-1KGEA
512
USD
BT225850-1KG
BT225850-250G
BT225850-50G
Tetrabutylammonium hydroxide 55% w/w water
Tetrabutylammonium hydroxide
CAS: 2052-49-5; EC No: 218-147-6; MDL No: MFCD00009425; RTECS: BS5425000
UN No: UN3267; Haz Class: 8; Packing Group: II
Liquid; Linear Formula: [CH3(CH2)3]4N(OH); Molecular Formula: C16H37NO; MW: 259.48
Boiling Point:
- RTECS: BS5425000
- Refractive index: 1.435
- Air Sensitive
- Liquid
Formula:
C₁₆H₃₇NO MW: 259.48 Boiling Pt: >100 °C Melting Pt: 29.85 Density: 0.995 g/mL |
MDL Number:
MFCD00009425 CAS Number: 2052-49-5 EINECS: 218-147-6 UN: 3267 ADR: 8 II |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...