1-Bromo-2-chloroethane ≥98%
Supplier: BeanTown Chemical
Synonyms:
Ethylene chlorobromide
225610-100G
225610-500G
BT225610-100GEA
49
USD
BT225610-100G
BT225610-500G
1-Bromo-2-chloroethane ≥98%
1-Bromo-2-chloroethane
CAS: 107-04-0; EC No: 203-456-0; MDL No: MFCD00000962; RTECS: KH6500000
UN No: UN2810; Haz Class: 6.1; Packing Group: III
Liquid; Molecular Formula: C2H4BrCl; MW: 143.41
Melting Point: -18°; Boiling Point: 106-107°
Density (g/mL): 1.723; Refractive Index: 1.488
Light Sensitive
- RTECS: KH6500000
- Refractive index: 1.488
- Light Sensitive
- Liquid
Formula:
BrH₂CCH₂Cl MW: 143.41 g/mol Boiling Pt: 107 Melting Pt: -17 Density: 1.723 g/mL |
MDL Number:
MFCD00000962 CAS Number: 107-04-0 EINECS: 203-456-0 UN: 2810 ADR: 6.1 III |
Specification Test Results
Assay | 98% |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...