Potassium isopropoxide ≥99%
Supplier: BeanTown Chemical
225590-100ML
225590-25ML
BT225590-100MLEA
264
USD
BT225590-100ML
BT225590-25ML
Potassium isopropoxide ≥99%
Potassium isopropoxide
CAS: 6831-82-9; EC No: 229-909-2; MDL No: MFCD00210641
UN No: UN3274; Haz Class: 3 (8); Packing Group: II
Liquid; Linear Formula: KOCH(CH3)2; Molecular Formula: C3H7KO; MW: 98.19
Air Sensitive, Moisture Sensitive
- Air Sensitive, Moisture Sensitive
- Liquid
Formula:
C₃H₇KO |
MDL Number:
MFCD00210641 CAS Number: 6831-82-9 EINECS: 229-909-2 ADR: 3 (8),II |
Specification Test Results
Assay | 99% |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...