Hydrochloric acid ≥33.0% ≥99.99999% (trace metals basis)

Supplier: BeanTown Chemical

225500-250ML 225500-25ML
BT225500-250MLEA 548.46 USD
BT225500-250ML BT225500-25ML
Hydrochloric acid ≥33.0% ≥99.99999% (trace metals basis)
Hydrochloric acid

CAS: 7647-01-0; EC No: 231-595-7; MDL No: MFCD00011324; RTECS: MW4025000 UN No: UN1789; Haz Class: 8; Packing Group: II Liquid, double distilled in quartz; Molecular Formula: HCl; MW: 36.46 Boiling Point: 100° Density (g/mL): 1.19


  • RTECS: MW4025000
  • Liquid, double distilled in quartz

Formula: HCl
Boiling Pt: >100 °C
Density: 1.19 g/mL
MDL Number: MFCD00011324
CAS Number: 7647-01-0
EINECS: 231-595-7
UN: 1789
ADR: 8,II

Order Now

Specification Test Results

Assay 99.99999%


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR