Hydroxylammonium chloride ≥99%

Supplier: BeanTown Chemical

Synonyms: Hydroxylamine hydrochlorideHydroxylamine HCl

225430-100G 225430-500G
BT225430-100GEA 31.91 USD
BT225430-100G BT225430-500G
Hydroxylammonium chloride ≥99%
Hydroxylammonium chloride

  • RTECS: NC3675000
  • Hygroscopic
  • Crystalline

Formula: NH₂OH·HCl
MW: 69.49 g/mol
Melting Pt: 155…157 °C (dec.)
Density: 1.67 g/mL
MDL Number: MFCD00051089
CAS Number: 5470-11-1
EINECS: 226-798-2
UN: 2923
ADR: 8,III

Order Now

Specification Test Results

Assay 99%

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR