Hydrochloric acid ≥36.5% ≥99.999% (trace metals basis)
Supplier: BeanTown Chemical
225305-1L
225305-2.5L
225305-250ML
BT225305-1LEA
211
USD
BT225305-1L
BT225305-2.5L
BT225305-250ML
Hydrochloric acid ≥36.5% ≥99.999% (trace metals basis)
Hydrochloric acid
CAS: 7647-01-0; EC No: 231-595-7; MDL No: MFCD00011324; RTECS: MW4025000
UN No: UN1789; Haz Class: 8; Packing Group: II
Liquid; Molecular Formula: HCl; MW: 36.46
Boiling Point:
- RTECS: MW4025000
- Liquid
Formula:
HCl Boiling Pt: >100 °C Density: 1.19 g/mL |
MDL Number:
MFCD00011324 CAS Number: 7647-01-0 EINECS: 231-595-7 UN: 1789 ADR: 8,II |
Specification Test Results
Assay | 99.999% |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...