Cyclohexanone ≥99%

Supplier: BeanTown Chemical

Synonyms: Pimelic Ketone

225300-500ML 225300-2.5L 225300-100ML
BT225300-2.5LEA 67.37 USD
BT225300-2.5L BT225300-100ML BT225300-500ML
Cyclohexanone ≥99%
Cyclohexanone

  • RTECS: GW1050000
  • Refractive index: 1.450
  • Liquid

Formula: C₆H₁₀(=O)
MW: 98.14 g/mol
Boiling Pt: 155 °C
Melting Pt: –47 °C
Density: 0.947 g/mL
Flash Pt: 44 °C (111 °F)
MDL Number: MFCD00001625
CAS Number: 108-94-1
EINECS: 203-631-1
UN: 1915
ADR: 3,III
https://us.vwr.com/stibo/web/std.lang.all/12/57/771257.jpg

Order Now

Specification Test Results

Assay 99%

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR