Tellurium ≥99.999% (trace metals basis), powder -60 mesh
Supplier: BeanTown Chemical
225265-10G
225265-250G
225265-50G
BT225265-10GEA
44.01
USD
BT225265-10G
BT225265-250G
BT225265-50G
Tellurium ≥99.999% (trace metals basis), powder -60 mesh
Tellurium
CAS: 13494-80-9; EC No: 236-813-4; MDL No: MFCD00134062; RTECS: WY2625000
UN No: UN3288; Haz Class: 6.1; Packing Group: III
Powder; Molecular Formula: Te; MW: 127.60
Melting Point: 450°; Boiling Point: 990°
Density (g/mL): 6.24
- RTECS: WY2625000
- Powder
Formula:
Te MW: 127.6 g/mol Boiling Pt: 990 Melting Pt: 450 Density: 6.24 g/mL |
MDL Number:
MFCD00134062 CAS Number: 13494-80-9 EINECS: 236-813-4 ADR: 6.1 III Merck Index: 12,09273 |
Specification Test Results
Assay | 99.999% |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...