Diethyl chlorophosphite ≥97%
Supplier: BeanTown Chemical
CAS: 589-57-1; EC No: 209-652-2; MDL No: MFCD00009074
UN No: UN2920; Haz Class: 8 (3); Packing Group: II
Liquid; Molecular Formula: C4H10ClO2P; MW: 156.55
Boiling Point: 153-155°; Flash point: 1°C (34°F)
Density (g/mL): 1.089; Refractive Index: 1.434
Moisture Sensitive
- Refractive index: 1.434
- Moisture Sensitive
- Liquid
Formula:
C₄H₁₀ClO₂P Boiling Pt: 153…155 °C Density: 1.089 g/mL Flash Pt: 1 °C (34 °F) |
MDL Number:
MFCD00009074 CAS Number: 589-57-1 EINECS: 209-652-2 UN: 2920 ADR: 8,II |
Specification Test Results
Assay | 97% |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...