Hydrochloric acid 34.0 - 37.5%
Supplier: BeanTown Chemical
CAS: 7647-01-0; EC No: 231-595-7; MDL No: MFCD00011324; RTECS: MW4025000
UN No: UN1789; Haz Class: 8; Packing Group: II
Liquid, single sub-boiling quartz distillation; Molecular Formula: HCl; MW: 36.46
Boiling Point:
- RTECS: MW4025000
- Liquid, single sub-boiling quartz distillation
Formula:
HCl Boiling Pt: >100 °C Density: 1.19 g/mL |
MDL Number:
MFCD00011324 CAS Number: 7647-01-0 EINECS: 231-595-7 UN: 1789 ADR: 8,II |
Specification Test Results
Assay | 34.0-37.5% |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...