Calcium peroxide

Supplier: BeanTown Chemical

224545-100G 224545-500G
BT224545-100GEA 81.15 USD
BT224545-100G BT224545-500G
Calcium peroxide
Calcium peroxide

CAS: 1305-79-9; EC No: 215-139-4; MDL No: MFCD00015988 UN No: UN1457; Haz Class: 5.1; Packing Group: II Powder; Molecular Formula: CaO2; MW: 72.08 Moisture Sensitive


  • Moisture Sensitive
  • Powder

Formula: CaO₂
MDL Number: MFCD00015988
CAS Number: 1305-79-9
EINECS: 215-139-4
UN: 1457
ADR: 5.1,II

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR