Chloroform ≥99.8% stabilised ACS for HPLC
Supplier: BeanTown Chemical
Synonyms:
Methane, trichloro-, Trichloroform, Methylidyne trichloride, Trichloromethane
Stabilization:
Stabilised with ethanol
224225-1L
224225-4L
224225-4X4L
BT224225-1LEA
55.84
USD
BT224225-1L
BT224225-4L
BT224225-4X4L
Chloroform ≥99.8% stabilised ACS for HPLC
Chloroform
CAS: 67-66-3; EC No: 200-663-8; MDL No: MFCD00000826; RTECS: FS9100000
UN No: UN1888; Haz Class: 6.1; Packing Group: III
Liquid; Molecular Formula: CHCl3; MW: 119.38
Melting Point: -63°; Boiling Point: 60.5-61.5°
Density (g/mL): 1.492; Refractive Index: 1.445
- RTECS: FS9100000
- Refractive index: 1.445
- Liquid
Formula:
CHCl₃ Boiling Pt: 60.5…61.5 °C Melting Pt: –63 °C Density: 1.492 g/mL |
MDL Number:
MFCD00000826 CAS Number: 67-66-3 EINECS: 200-663-8 UN: 1888 ADR: 6.1,III |
|
Specification Test Results
Assay | 99.8% min. |
Color (APHA) | ≤10 |
Residue after evaporation | ≤0.001% |
Acetone and aldehyde | Passes test |
Acid and Chloride | Passes test |
Free chloride (Cl) | Passes test |
Lead (Pb) | ≤0.05ppm |
Substances darkened by H2SO4 | Passes test |
Suitability for use in dithizone test | Passes test |
Ethanol stabilizer | 0.5-1.5% |
Water | ≤0.03% |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...