1-Methylnaphthalene ≥96%

Supplier: BeanTown Chemical

224060-100G 224060-25G 224060-500G
BT224060-100GEA 26.11 USD
BT224060-100G BT224060-25G BT224060-500G
1-Methylnaphthalene ≥96%
1-Methylnaphthalene

CAS: 90-12-0; EC No: 201-966-8; MDL No: MFCD00004034; RTECS: QJ9630000 UN No: UN3082; Haz Class: 9; Packing Group: III Liquid; Molecular Formula: C11H10; MW: 142.20 Melting Point: -22°; Boiling Point: 240-243°; Flash point: 82°C (180°F) Density (g/mL): 1.001; Refractive Index: 1.615


  • RTECS: QJ9630000
  • Refractive index: 1.615
  • Liquid

Formula: C₁₁H₁₀
Boiling Pt: 240…243 °C
Melting Pt: –22 °C
Density: 1.001 g/mL
Flash Pt: 82 °C (180 °F)
MDL Number: MFCD00004034
CAS Number: 90-12-0
EINECS: 201-966-8
UN: 3082
ADR: 9,III

Order Now

Specification Test Results

Assay 96%


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR