(S)-(+)-2-Amino-1-phenylethanol ≥97%

Supplier: BeanTown Chemical

224000-1G 224000-5G
BT224000-1GEA 75 USD
BT224000-1G BT224000-5G
(S)-(+)-2-Amino-1-phenylethanol ≥97%
(S)-(+)-2-Amino-1-phenylethanol

CAS: 56613-81-1; MDL No: MFCD00239405 UN No: UN3259; Haz Class: 8; Packing Group: III Powder; Molecular Formula: C8H11NO ; MW: 137.18 Melting Point: 57-59°; Boiling Point: 160°/17 mmHg Optical Rotation: [α]20/D +43±2°, c = 2% in ethanol Air Sensitive


  • Optical rotation: [α]20/D +43±2°, c = 2% in ethanol
  • Air Sensitive
  • Powder

Formula: C₈H₁₁NO
MW: 137.182 g/mol
Boiling Pt: 160 °C/17 mmHg
Melting Pt: 57…59 °C
MDL Number: MFCD00239405
CAS Number: 56613-81-1
ADR: 8 III

Order Now

Specification Test Results

Assay 97%


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR