Zinc chloride, anhydrous ≥99.95% (trace metals basis)
Supplier: BeanTown Chemical
CAS: 7646-85-7; EC No: 231-592-0; MDL No: MFCD00011295; RTECS: ZH1400000
UN No: UN2331; Haz Class: 8; Packing Group: III
Crystalline/Powder; Linear Formula: ZnCl2; MW: 136.29
Melting Point: 293°; Boiling Point: 732°
Density (g/mL): 2.907
Hygroscopic
- RTECS: ZH1400000
- Hygroscopic
- Crystalline/Powder
Formula:
ZnCl₂ Boiling Pt: 732 °C Melting Pt: 293 °C Density: 2.907 g/mL |
MDL Number:
MFCD00011295 CAS Number: 7646-85-7 EINECS: 231-592-0 UN: 2331 ADR: 8,III |
Specification Test Results
Assay | 99.95% |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...