Heptafluorobutyric acid ≥99%

Supplier: BeanTown Chemical

223730-100G 223730-25G 223730-500G
BT223730-25GEA 43.99 USD
BT223730-25G BT223730-500G BT223730-100G
Heptafluorobutyric acid ≥99%
Heptafluorobutyric acid

  • RTECS: ET4025000
  • Light Sensitive
  • Liquid

Formula: C₄HF₇O₂
MW: 214.04 g/mol
Boiling Pt: 120 °C/755 mmHg
Density: 1.645 g/mL
MDL Number: MFCD00004171
CAS Number: 375-22-4
EINECS: 206-786-3
UN: 3265
ADR: 8,II

Order Now

Specification Test Results

Assay 99%

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR