Titanium(IV) isopropoxide ≥97%

Supplier: BeanTown Chemical

Synonyms: Titanium (IV) isopropoxideTitanium tetraisopropanolate

223400-100G 223400-500G
BT223400-500GEA 70.99 USD
BT223400-500G BT223400-100G
Titanium(IV) isopropoxide ≥97%
Titanium (IV) isopropoxide

  • RTECS: NT8060000
  • Refractive index: 1.464
  • Moisture Sensitive
  • Liquid

Formula: C₁₂H₂₈O₄Ti
MW: 284.23 g/mol
Boiling Pt: 232 °C
Melting Pt: 14…17 °C
Density: 0.955 g/mL
Flash Pt: 45 °C (113 °F)
MDL Number: MFCD00008871
CAS Number: 546-68-9
EINECS: 208-909-6
UN: 2413
ADR: 3,III

Order Now

Specification Test Results

Assay 97%

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR