Methyl orange ACS

Supplier: BeanTown Chemical

Synonyms: 4-((4-(Dimethylamino)phenyl)diazenyl)benzenesulphonic acid, sodium saltMethyl orange sodium saltHelianthine4-((4-(Dimethylamino)phenyl)diazenyl)benzenesulfonic acid, sodium salt

223275-100G 223275-5G 223275-25G
BT223275-5GEA 12.55 USD
BT223275-5G BT223275-25G BT223275-100G
Methyl orange ACS
Methyl orange

  • RTECS: DB6327000
  • Crystalline/Powder

Formula: C₁₄H₁₄N₃NaO₃S
MW: 327.34 g/mol
Melting Pt: >300 °C
MDL Number: MFCD00007502
CAS Number: 547-58-0
EINECS: 208-925-3
UN: 3143
ADR: 6.1,III

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR