2-(Dimethylamino)-ethanol ≥99%

Supplier: BeanTown Chemical

Synonyms: 2-DimethylaminoethanolN,N-DimethylethanolamineDMAE

223190-250ML 223190-1L
BT223190-250MLEA 24.28 USD
BT223190-250ML BT223190-1L
2-(Dimethylamino)-ethanol ≥99%
2-Dimethylaminoethanol

  • RTECS: KK6125000
  • Refractive index: 1.429
  • Hygroscopic
  • Liquid

Formula: C₄H₁₁NO
MW: 89.14 g/mol
Boiling Pt: 134…136 °C
Melting Pt: –70 °C
Density: 0.886 g/mL
Flash Pt: 39 °C (102 °F)
MDL Number: MFCD00002846
CAS Number: 108-01-0
EINECS: 203-542-8
UN: 2051
ADR: 8,II

Order Now

Specification Test Results

Assay 99%

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR