Indium antimonide ≥99.99%

Supplier: BeanTown Chemical

223090-1G 223090-5G
BT223090-1GEA 23 USD
BT223090-1G BT223090-5G
Indium antimonide ≥99.99%
Indium antimonide

CAS: 1312-41-0; EC No: 215-192-3; MDL No: MFCD00016146 UN No: UN1549; Haz Class: 6.1; Packing Group: III Pieces; Linear Formula: InSb; MW: 236.57 Melting Point: 535°


  • Pieces

Formula: InSb
MW: 236.58
Melting Pt: 527
Density: 5.78
MDL Number: MFCD00016146
CAS Number: 1312-41-0
EINECS: 215-192-3
UN: 1549
ADR: 6.1 III
Merck Index: 13,04972

Order Now

Specification Test Results

Assay 99.99%


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR