(3-Mercaptopropyl)trimethoxysilane ≥95%

Supplier: BeanTown Chemical

Synonyms: 3-(Trimethoxysilyl)propane-1-thiolTrimethoxy(3-mercaptopropyl)silane3-(Trimethoxysilyl)-1-propanethiol

222870-25G 222870-100G 222870-500G
BT222870-25GEA 29.76 USD
BT222870-25G BT222870-100G BT222870-500G
(3-Mercaptopropyl)trimethoxysilane ≥95%
(3-Mercaptopropyl)trimethoxysilane

  • RTECS: TZ7800000
  • Refractive index: 1.443
  • Air Sensitive, Moisture Sensitive
  • Liquid

Formula: HSH₂CCH₂CH₂Si(OCH₃)₃
MW: 196.34 g/mol
Boiling Pt: 213…215 °C
Density: 1.05 g/mL
Flash Pt: 96 °C (205 °F)
MDL Number: MFCD00004901
CAS Number: 4420-74-0
EINECS: 224-588-5
UN: 3082
ADR: 9,III

Order Now

Specification Test Results

Assay 95%

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR