Periodic acid 99.0-101.0% ACS

Supplier: BeanTown Chemical

Synonyms: Orthoperiodic acidortho-Periodic acid

222655-50G
BT222655-50GEA 108.74 USD
BT222655-50G
Periodic acid 99.0-101.0% ACS
Periodic acid

  • RTECS: NN1269700
  • Hygroscopic, Light Sensitive
  • Crystalline

Formula: H₅IO₆
MW: 227.94 g/mol
Melting Pt: 122 °C
MDL Number: MFCD00011344
CAS Number: 10450-60-9
EINECS: 233-937-0
UN: 3085
ADR: 5.1,II
https://us.vwr.com/stibo/web/std.lang.all/90/77/779077.jpg

Order Now

Specification Test Results

Assay 99.0-101.0%
Insoluble matter ≤0.01%
Residue after ignition ≤0.01%
Other halogens ≤0.01%
Sulfate (SO4) ≤0.01%
Heavy metals (as Pb) ≤005%
Iron (Fe) ≤003%

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR