Diethyl carbonate ≥99%
Supplier: BeanTown Chemical
222000-10L
222000-2.5L
222000-500ML
BT222000-10LEA
426.91
USD
BT222000-10L
BT222000-2.5L
BT222000-500ML
Diethyl carbonate ≥99%
Diethyl carbonate
CAS: 105-58-8; EC No: 203-311-1; MDL No: MFCD00009107; RTECS: FF9800000
UN No: UN2366; Haz Class: 3; Packing Group: III
Liquid; Linear Formula: (CH3CH2O)2CO; Molecular Formula: C5H10O5; MW: 118.13
Melting Point: -43°; Boiling Point: 126-128°; Flash point: 25°C (77°F)
Density (g/mL): 0.975; Refractive Index: 1.384
Moisture Sensitive
- RTECS: FF9800000
- Refractive index: 1.384
- Moisture Sensitive
- Liquid
Formula:
C₅H₁₀O₃ MW: 118.13 g/mol Boiling Pt: 127 Melting Pt: -43 Density: 0.975 g/mL Flash Pt: 25 °C (77 °F) |
MDL Number:
MFCD00009107 CAS Number: 105-58-8 EINECS: 203-311-1 UN: 2366 ADR: 3 III Merck Index: 13,03815 |
Specification Test Results
Assay | 99% |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...