Chloroform-[D₁] 99,8 isotopic contains 1% TMS NMR

Supplier: BeanTown Chemical

Synonyms: Trichloromethane-D1, Chloroform-[²H₁], Chloroform-D

221910-100G 221910-25G 221910-500G
BT221910-100GEA 85.42 USD
BT221910-100G BT221910-25G BT221910-500G
Chloroform-[D₁] 99,8 isotopic contains 1% TMS NMR
Chloroform-[D1]

CAS: 865-49-6; EC No: 212-742-4; MDL No: MFCD00000827 UN No: UN1888; Haz Class: 6.1; Packing Group: III Liquid; Molecular Formula: CDCl3; MW: 120.38 Melting Point: -64°; Boiling Point: 60.9° Density (g/mL): 1.5; Refractive Index: 1.444 Light Sensitive, Moisture Sensitive


  • Refractive index: 1.444
  • Light Sensitive, Moisture Sensitive
  • Liquid

Formula: CDCl₃
MW: 120.38 g/mol
Boiling Pt: 60.8
Melting Pt: -64.1
Density: 1.5 g/mL
MDL Number: MFCD00000827
CAS Number: 865-49-6
EINECS: 212-742-4
UN: 1888
ADR: 6.1 III

Order Now

Specification Test Results

Assay 99.8%


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR