Platinum ≥99.9% (trace metals basis), Powder -325 mesh
Supplier: BeanTown Chemical
221655-1G
221655-250MG
221655-5G
BT221655-1GEA
311
USD
BT221655-1G
BT221655-250MG
BT221655-5G
Platinum ≥99.9% (trace metals basis), Powder -325 mesh
Platinum
CAS: 7440-06-4; MDL No: MFCD00011179; RTECS: TP2160000
UN No: UN3089; Haz Class: 4.1; Packing Group: III
Powder; Molecular Formula: Pt; MW: 195.08
Melting Point: 1772°; Boiling Point: 3827°
Density (g/mL): 21.45
- RTECS: TP2160000
- Powder
Formula:
Pt Boiling Pt: 3827 °C Melting Pt: 1772 °C Density: 21.45 g/mL |
MDL Number:
MFCD00011179 CAS Number: 7440-06-4 UN: 3089 ADR: 4.1,III |
Specification Test Results
Assay | 99.9% |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...