Potassium dicyanoargentate ≥99.9%

Supplier: BeanTown Chemical

Synonyms: Potassium silver cyanide

221480-100G 221480-25G 221480-500G
BT221480-100GEA 328 USD
BT221480-100G BT221480-25G BT221480-500G
Potassium dicyanoargentate ≥99.9%
Potassium dicyanoargentate

CAS: 506-61-6; EC No: 208-047-0; MDL No: MFCD00036297; RTECS: TT5775000 UN No: UN1588; Haz Class: 6.1; Packing Group: I Powder; Linear Formula: KAg(CN)2; MW: 199.01 Density (g/mL): 2.36 Light Sensitive


  • RTECS: TT5775000
  • Light Sensitive
  • Powder

Formula: C₂AgKN₂
Density: 2.36 g/mL
MDL Number: MFCD00036297
CAS Number: 506-61-6
EINECS: 208-047-0
UN: 1588
ADR: 6.1,I

Order Now

Specification Test Results

Assay 99.9%


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR