2-Methylcyclohexanone ≥98%
Supplier: BeanTown Chemical
221350-100ML
221350-500ML
221350-2.5L
BT221350-100MLEA
98.83
USD
BT221350-100ML
BT221350-500ML
BT221350-2.5L
2-Methylcyclohexanone ≥98%
2-Methylcyclohexanone
- RTECS: GW1750000
- Refractive index: 1.448
- Liquid
Formula:
C₇H₁₂O MW: 112.17 g/mol Boiling Pt: 162…163 °C Density: 0.924 g/mL Flash Pt: 48 °C (118 °F) |
MDL Number:
MFCD00001635 CAS Number: 583-60-8 EINECS: 209-513-6 UN: 2297 ADR: 3,III |
Specification Test Results
Assay | 98% |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...