Potassium dichromate ≥99%
Supplier: BeanTown Chemical
221150-100G
221150-2.5KG
221150-500G
BT221150-100GEA
53.77
USD
BT221150-100G
BT221150-2.5KG
BT221150-500G
Potassium dichromate ≥99%
Potassium dichromate
CAS: 7778-50-9; EC No: 231-906-6; MDL No: MFCD00011367; RTECS: HX7680000
UN No: UN3087; Haz Class: 5.1 (6.1); Packing Group: II
Crystalline; Linear Formula: K2Cr2O7; MW: 294.18
Melting Point: 398°
Density (g/mL): 2.68
- RTECS: HX7680000
- Crystalline
Formula:
K₂Cr₂O₇ Melting Pt: 398 °C Density: 2.68 g/mL |
MDL Number:
MFCD00011367 CAS Number: 7778-50-9 EINECS: 231-906-6 UN: 3087 ADR: 5.1,II |
Specification Test Results
Assay | 99% |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...