3-Cyclohexene-1-carboxylic acid ≥98%

Supplier: BeanTown Chemical

221120-25G 221120-5G
BT221120-25GEA 288 USD
BT221120-25G BT221120-5G
3-Cyclohexene-1-carboxylic acid ≥98%
3-Cyclohexene-1-carboxylic acid

CAS: 4771-80-6; EC No: 225-314-7; MDL No: MFCD00013781; RTECS: GW3675000 UN No: UN3265; Haz Class: 8; Packing Group: III Liquid; Molecular Formula: C7H10O2 ; MW: 126.15 Melting Point: 17°; Boiling Point: 130-133°/4 mmHg; Flash point: 110°C (230°F) Density (g/mL): 1.081; Refractive Index: 1.48


  • RTECS: GW3675000
  • Refractive index: 1.48
  • Liquid

Formula: C₇H₁₀O₂
Boiling Pt: 130…133 °C/4 mmHg
Melting Pt: 17 °C
Density: 1.081 g/mL
Flash Pt: 110 °C (230 °F)
MDL Number: MFCD00013781
CAS Number: 4771-80-6
EINECS: 225-314-7
ADR: 8,III

Order Now

Specification Test Results

Assay 98%


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR