Ammonium nitrate ≥95% ACS

Supplier: BeanTown Chemical

221040-2KG 221040-500G
BT221040-2KGEA 140.51 USD
BT221040-2KG BT221040-500G
Ammonium nitrate ≥95% ACS
Ammonium nitrate

CAS: 6484-52-2; EC No: 229-347-8; MDL No: MFCD00011425; RTECS: BR9050000 UN No: UN1942; Haz Class: 5.1; Packing Group: III Granular; Linear Formula: NH4NO3; MW: 80.04 Melting Point: 169°; Boiling Point: 210°


  • RTECS: BR9050000
  • Granular

Formula: NH₄NO₃
MW: 80.04 g/mol
Boiling Pt: 230
Melting Pt: 169
MDL Number: MFCD00011425
CAS Number: 6484-52-2
EINECS: 229-347-8
UN: 1942
ADR: 5.1 III
Merck Index: 13,00538

Order Now

Specification Test Results

Assay 95% min.


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR