1-Ethoxy-1-(tributylstannyl)ethene
Supplier: BeanTown Chemical
Synonyms:
Tri-n-butyl(1-ethoxyvinyl)tin
, 1-Ethoxyvinyltri-n-butyltin
, Tributyl(1-ethoxyvinyl)tin
220780-25G
220780-1G
220780-5G
BT220780-5GEA
215
USD
BT220780-5G
BT220780-1G
BT220780-25G
1-Ethoxy-1-(tributylstannyl)ethene
1-Ethoxy-1-(tributylstannyl)ethene
- Refractive index: 1.476
- Liquid
Formula:
C₁₆H₃₄OSn MW: 361.15 g/mol Boiling Pt: 85…86 °C/0.1 mmHg Density: 1.069 g/mL Flash Pt: >110 °C (>230 °F) |
MDL Number:
MFCD00010240 CAS Number: 97674-02-7 ADR: 6.1,III |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...