Ethylphosphonic acid ≥98%
Supplier: BeanTown Chemical
220445-1G
220445-25G
220445-5G
BT220445-1GEA
66.92
USD
BT220445-1G
BT220445-25G
BT220445-5G
Ethylphosphonic acid ≥98%
Ethylphosphonic acid
CAS: 6779-09-5; EC No: 229-839-2; MDL No: MFCD00002141
UN No: UN3261; Haz Class: 8; Packing Group: II
Powder; Molecular Formula: C2H7O3P; MW: 110.06
Melting Point: 61-63°
Hygroscopic
- Hygroscopic
- Powder
Formula:
C₂H₇O₃P MW: 110.05 g/mol Melting Pt: 61…63 °C |
MDL Number:
MFCD00002141 CAS Number: 6779-09-5 EINECS: 229-839-2 UN: 3261 ADR: 8 II |
Specification Test Results
Assay | 98% |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...