1H-Tetrazole 0.45M in acetonitrile ≥98%
Supplier: BeanTown Chemical
Synonyms:
Tetrazole
220365-200ML
220365-450ML
220365-45ML
BT220365-200MLEA
305.95
USD
BT220365-200ML
BT220365-450ML
BT220365-45ML
1H-Tetrazole 0.45M in acetonitrile ≥98%
1H-Tetrazole
CAS: 288-94-8; EC No: 206-023-4; MDL No: MFCD00005247
UN No: UN1648; Haz Class: 3; Packing Group: II
Liquid; Molecular Formula: CH2N4 ; MW: 70.05
Flash point: 5°C (41°F)
Density (g/mL): 0.798
- Liquid
Formula:
CH₂N₄ Density: 0.798 g/mL Flash Pt: 5 °C (41 °F) |
MDL Number:
MFCD00005247 CAS Number: 288-94-8 EINECS: 206-023-4 ADR: 3,II |
Specification Test Results
Assay | 98% |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...