Hexafluorophosphoric acid 60% (w/w) in water
Supplier: BeanTown Chemical
220290-100ML
220290-25ML
BT220290-100MLEA
56.06
USD
BT220290-100ML
BT220290-25ML
BT220290-500ML
Hexafluorophosphoric acid 60% (w/w) in water
Hexafluorophosphoric acid
CAS: 16940-81-1; EC No: 241-006-5; MDL No: MFCD00011350; RTECS: SY7115000
UN No: UN1782; Haz Class: 8; Packing Group: II
Liquid; Linear Formula: HPF6; MW: 145.97
Density (g/mL): 1.651
- RTECS: SY7115000
- Liquid
Formula:
HF₆P Density: 1.651 g/mL |
MDL Number:
MFCD00011350 CAS Number: 16940-81-1 EINECS: 241-006-5 UN: 1782 ADR: 8,II |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...