Chromium ≥99% (trace metals basis), Powder APS <10 µm
Supplier: BeanTown Chemical
220075-1KG
220075-250G
220075-50G
BT220075-1KGEA
467
USD
BT220075-1KG
BT220075-250G
BT220075-50G
Chromium ≥99% (trace metals basis), Powder APS <10 µm
Chromium
CAS: 7440-47-3; EC No: 231-157-5; MDL No: MFCD00010944; RTECS: GB4200000
UN No: UN3089; Haz Class: 4.1; Packing Group: III
Powder; Molecular Formula: Cr; MW: 52.00
Melting Point: 1857°; Boiling Point: 2672°
Density (g/mL): 7.14
- RTECS: GB4200000
- Powder
Formula:
Cr Boiling Pt: 2672 °C Melting Pt: 1857 °C Density: 7.14 g/mL |
MDL Number:
MFCD00010944 CAS Number: 7440-47-3 EINECS: 231-157-5 UN: 3089 ADR: 4.1,III |
Specification Test Results
Assay | 99% |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...