Pyrogallol ≥99.0% ACS
Supplier: BeanTown Chemical
Synonyms:
Benzene-1,2,3-triol
, 1,2,3-Trihydroxybenzene
220065-50G
220065-10G
220065-250G
BT220065-250GEA
209.8
USD
BT220065-250G
BT220065-50G
BT220065-10G
Pyrogallol ≥99.0% ACS
Pyrogallol
- RTECS: UX2800000
- Air Sensitive, Light Sensitive
- Crystalline/Powder
Formula:
C₆H₆O₃ MW: 126.11 g/mol Boiling Pt: 309 °C Melting Pt: 131…135 °C Density: 1.45 g/mL |
MDL Number:
MFCD00002192 CAS Number: 87-66-1 EINECS: 201-762-9 UN: 2811 ADR: 6.1,III |
Specification Test Results
Assay | 99.0% min. |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...