Sodium ethoxide 21% (w/w) in ethanol

Supplier: BeanTown Chemical

Synonyms: Sodium ethylate, Sodium ethanolate

219925-100ML 219925-2.5L 219925-500ML
BT219925-100MLEA 27.98 USD
BT219925-100ML BT219925-2.5L BT219925-500ML
Sodium ethoxide 21% (w/w) in ethanol
Sodium ethoxide

CAS: 141-52-6; EC No: 205-487-5; MDL No: MFCD00012417 UN No: UN3274; Haz Class: 3 (8); Packing Group: II Liquid; Linear Formula: CH3CH2ONa; MW: 68.05 Boiling Point: 91°; Flash point: 9°C (48°F) Density (g/mL): 0.868; Refractive Index: 1.385 Air Sensitive, Moisture Sensitive


  • Refractive index: 1.385
  • Air Sensitive, Moisture Sensitive
  • Liquid

Formula: C₂H₅NaO
MW: 68.05 g/mol
Boiling Pt: 91 °C
Melting Pt: 260
Density: 0.868 g/mL
Flash Pt: 9 °C (48 °F)
MDL Number: MFCD00012417
CAS Number: 141-52-6
EINECS: 205-487-5
UN: 3274
ADR: 3 II

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR