Sodium nitrate ≥99.999%
Supplier: BeanTown Chemical
219265-100G
219265-25G
219265-5G
BT219265-100GEA
612
USD
BT219265-100G
BT219265-25G
BT219265-5G
Sodium nitrate ≥99.999%
Sodium nitrate
CAS: 7631-99-4; EC No: 231-554-3; MDL No: MFCD00011119; RTECS: WC5600000
UN No: UN1498; Haz Class: 5.1; Packing Group: III
Solid; Linear Formula: NaNO3; MW: 84.99
Melting Point: 306° (decomposes); Boiling Point: 380°
Density (g/mL): 2.261
- RTECS: WC5600000
- Solid
Formula:
NaNO₃ Boiling Pt: 380 °C Melting Pt: 306 °C (dec.) Density: 2.261 g/mL |
MDL Number:
MFCD00011119 CAS Number: 7631-99-4 EINECS: 231-554-3 UN: 1498 ADR: 5.1,III |
Specification Test Results
Assay | 99.999% |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...