Dimethyl carbonate ≥99%
Supplier: BeanTown Chemical
219220-100G
219220-2.5KG
219220-500G
BT219220-100GEA
22.04
USD
BT219220-100G
BT219220-2.5KG
BT219220-500G
Dimethyl carbonate ≥99%
Dimethyl carbonate
CAS: 616-38-6; EC No: 210-478-4; MDL No: MFCD00008420; RTECS: FG0450000
UN No: UN1161; Haz Class: 3; Packing Group: II
Liquid; Linear Formula: (CH3O)2CO; MW: 90.08
Melting Point: 2-4°; Boiling Point: 90°; Flash point: 16°C (61°F)
Density (g/mL): 1.069; Refractive Index: 1.368
Moisture Sensitive
- RTECS: FG0450000
- Refractive index: 1.368
- Moisture Sensitive
- Liquid
Formula:
C₃H₆O₃ Boiling Pt: 90 °C Melting Pt: 2…4 °C Density: 1.069 g/mL Flash Pt: 16 °C (61 °F) |
MDL Number:
MFCD00008420 CAS Number: 616-38-6 EINECS: 210-478-4 UN: 1161 ADR: 3,II |
Specification Test Results
Assay | 99% |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...