Chloramine-T (sodium salt) trihydrate 98.0-103.0% ACS

Supplier: BeanTown Chemical

Synonyms: Tosylchloramide sodium trihydrate, Chlorine bleaching solution, Chloramine T trihydrate, Chloramine T sodium salt trihydrate, N-Chloro-4-toluenesulfonamide sodium salt, Sodium chloro(tosyl)amide trihydrate, N-Chloro-4-toluenesulphonamide sodium salt

219130-100G 219130-500G
BT219130-100GEA 40.56 USD
BT219130-100G BT219130-500G
Chloramine-T (sodium salt) trihydrate 98.0-103.0% ACS
Chloramine T (sodium salt) trihydrate

CAS: 7080-50-4; EC No: 204-854-7; MDL No: MFCD00149066; RTECS: XT5616800 UN No: UN3263; Haz Class: 8; Packing Group: III Crystalline; Molecular Formula: C7H7ClNNaO2S·3H2O; MW: 281.69 Melting Point: 167-170°; Flash point: 192°C (378°F) Moisture Sensitive


  • RTECS: XT5616800
  • Moisture Sensitive
  • Crystalline

Formula: CH₃C₆H₄SO₂NClNa·3H₂O
MW: 281.69 g/mol
Melting Pt: 167…170 °C
Flash Pt: 192 °C (378 °F)
MDL Number: MFCD00149066
CAS Number: 7080-50-4
EINECS: 204-854-7
UN: 3263
ADR: 8,III

Order Now

Specification Test Results

Assay 98.0-103.0%

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR