Titanium ≥90% (trace metals basis), powder <20 µm, Technical Grade
Supplier: BeanTown Chemical
219085-100G
219085-25G
219085-5G
BT219085-100GEA
327
USD
BT219085-100G
BT219085-25G
BT219085-5G
Titanium ≥90% (trace metals basis), powder <20 µm, Technical Grade
Titanium
CAS: 7440-32-6; EC No: 231-142-3; MDL No: MFCD00011264; RTECS: XR1700000
UN No: UN1352; Haz Class: 4.1; Packing Group: II
Powder, packaged in water; Molecular Formula: Ti; MW: 47.87
Melting Point: 1660°; Boiling Point: 3287°
Density (g/mL): 4.5
- RTECS: XR1700000
- Powder, packaged in water
Formula:
Ti MW: 47.88 g/mol Boiling Pt: 3262 Melting Pt: 1660 Density: 4.5 g/mL |
MDL Number:
MFCD00011264 CAS Number: 7440-32-6 EINECS: 231-142-3 UN: 1352 ADR: 4.2 II Merck Index: 12,09610 |
Specification Test Results
Assay | 90% |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...