Iodine 99,5, powder_crystalline
Supplier: BeanTown Chemical
219030-100G
219030-250G
219030-25G
BT219030-100GEA
68.1
USD
BT219030-100G
BT219030-250G
BT219030-25G
Iodine 99,5, powder_crystalline
Iodine
CAS: 7553-56-2; EC No: 231-442-4; MDL No: MFCD00011355; RTECS: NN1575000
UN No: UN3495; Haz Class: 8 (6.1); Packing Group: III
Solid; Molecular Formula: I2; MW: 253.81
Melting Point: 113°; Boiling Point: 184°
Density (g/mL): 4.94
- RTECS: NN1575000
- Solid
Formula:
I₂ MW: 253.81 g/mol Boiling Pt: 184.3 Melting Pt: 114 Density: 4.94 g/mL |
MDL Number:
MFCD00011355 CAS Number: 7553-56-2 EINECS: 231-442-4 UN: 3495 ADR: 8 III Merck Index: 13,05036 |
Specification Test Results
Assay | 99.5% |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...