Trichloroacetonitrile ≥98%

Supplier: BeanTown Chemical

218965-1KG 218965-250G 218965-50G
BT218965-1KGEA 266 USD
BT218965-1KG BT218965-250G BT218965-50G
Trichloroacetonitrile ≥98%
Trichloroacetonitrile

CAS: 545-06-2; EC No: 208-885-7; MDL No: MFCD00001842; RTECS: AM2450000 UN No: UN3276; Haz Class: 6.1; Packing Group: III Liquid; Linear Formula: CCl3CN; MW: 144.39 Melting Point: -44°; Boiling Point: 85-86°; Flash point: 195°C (383°F) Density (g/mL): 1.44; Refractive Index: 1.441


  • RTECS: AM2450000
  • Refractive index: 1.441
  • Liquid

Formula: C₂Cl₃N
MW: 144.39 g/mol
Boiling Pt: 85.7
Melting Pt: -44
Density: 1.44 g/mL
Flash Pt: 195 °C (383 °F)
MDL Number: MFCD00001842
CAS Number: 545-06-2
EINECS: 208-885-7
UN: 3276
ADR: 6.1 III
Merck Index: 12,09757

Order Now

Specification Test Results

Assay 98%


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR